Tantalum pentachloride 99.999%

Supplier: Apollo Scientific
Danger

IN3406-100G IN3406-250G IN3406-25G
APOSIN3406-100GEA 502 EUR
APOSIN3406-100G APOSIN3406-250G APOSIN3406-25G
Tantalum pentachloride 99.999%
Tantalum pentachloride
Formula: Cl₅Ta
MW: 358.21 g/mol
Boiling Pt: 242 °C
Melting Pt: 216 °C
Density: 3.68 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011253
CAS Number: 7721-01-9
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR