Tellurium 99.999+%, lumps
Supplier: Apollo Scientific
Danger
IN3433-100G IN3433-500G
APOSIN3433-100GEA 290 EUR
APOSIN3433-100G APOSIN3433-500G
Tellurium 99.999+%, lumps
Tellurium
Formula: Te
MW: 127.6 g/mol
Boiling Pt: 990 °C
Melting Pt: 449.5 °C
Density: 6.24 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134062
CAS Number: 13494-80-9
UN: 3288
ADR: 6.1,III