Tellurium dioxide 99.99%
Supplier: Apollo Scientific
Warning
IN3446-100G IN3446-25G
APOSIN3446-100GEA 545 EUR
APOSIN3446-100G APOSIN3446-25G
Tellurium dioxide 99.99%
Tellurium dioxide
Formula: O₂Te
MW: 159.6 g/mol
Boiling Pt: 1245 °C
Melting Pt: 733 °C
Density: 5.8 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011263
CAS Number: 7446-07-3
EINECS: 231-193-1
Merck Index: 14,09123