Tin(IV) oxide 99.9+%
Supplier: Apollo Scientific
Synonyms: Tin(iv)oxide, Stannic Oxide, Tin dioxide
IN3673-100G IN3673-500G IN3673-1KG
APOSIN3673-100GEA 87 EUR
APOSIN3673-100G APOSIN3673-500G APOSIN3673-1KG
Tin(IV) oxide 99.9+%
Tin (IV) oxide
Formula: O₂Sn
Boiling Pt: 1800…900 °C
Melting Pt: 1630 °C
Density: 6.95 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011244
CAS Number: 18282-10-5