Ytterbium(III) oxide 99,9%
Supplier: Apollo Scientific
Warning
Synonyms: Diytterbium trioxide, Ytterbium oxide
IN3811-10G IN3811-50G
APOSIN3811-10GEA 72.5 EUR
APOSIN3811-10G APOSIN3811-50G
Ytterbium(III) oxide 99,9%
Ytterbium (III) oxide
Formula: O₃Yb₂
MW: 394.08 g/mol
Melting Pt: 2227 °C
Density: 9.17 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011290
CAS Number: 1314-37-0
EINECS: 215-234-0