Diyttrium trioxide 99.999%
Supplier: Apollo Scientific
Warning
IN3865-100G IN3865-25G
APOSIN3865-100GEA 416 EUR
APOSIN3865-100G APOSIN3865-25G
Diyttrium trioxide 99.999%
Diyttrium trioxide
Formula: Y₂O₃
Boiling Pt: 4300 °C
Melting Pt: 2410 °C
Density: 4.84 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011473
CAS Number: 1314-36-9
EINECS: 215-233-5