2-(4-Aminophenyl)ethanol
Supplier: Apollo Scientific
Warning
Synonyms: 4-Aminophenethyl alcohol
OR0014-100G OR0014-25G OR0014-500G
APOSOR0014-100GEA 71.2 EUR
APOSOR0014-100G APOSOR0014-25G APOSOR0014-500G
2-(4-Aminophenyl)ethanol
2-(4-Aminophenyl)ethanol
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 255 °C
Melting Pt: 107…110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007922
CAS Number: 104-10-9
EINECS: 203-174-8