2-Aminopyridine 99%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Pyridylamine, Pyridin-2-amine
OR0015-1KG OR0015-100G OR0015-500G
APOSOR0015-1KGEA 108 EUR
APOSOR0015-1KG APOSOR0015-100G APOSOR0015-500G
2-Aminopyridine 99%
2-Aminopyridine
Formula: C₅H₆N₂
MW: 94.12 g/mol
Boiling Pt: 204…211 °C
Melting Pt: 54…60 °C
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00006312
CAS Number: 504-29-0
EINECS: 207-988-4
UN: 2671
ADR: 6.1 II
Merck Index: 13,00472