5-Chloro-1-indanone
Supplier: Apollo Scientific
Warning
OR0020-25G OR0020-5G
APOSOR0020-25GEA 24.8 EUR
APOSOR0020-25G APOSOR0020-5G
5-Chloro-1-indanone
5-Chloro-1-indanone
Formula: C₉H₇ClO
MW: 166.61 g/mol
Boiling Pt: 124…125 °C/3mmHg
Melting Pt: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00041456
CAS Number: 42348-86-7