1-Chloro-4-iodobenzene
Supplier: Apollo Scientific
Warning
OR0022-100G OR0022-1KG OR0022-500G
APOSOR0022-100GEA 29 EUR
APOSOR0022-100G APOSOR0022-1KG APOSOR0022-500G
1-Chloro-4-iodobenzene
1-Chloro-4-iodobenzene
Formula: C₆H₄ClI
MW: 238.45 g/mol
Boiling Pt: 226…227 °C
Melting Pt: 52…56 °C
Density: 1.886
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00001053
CAS Number: 637-87-6
EINECS: 211-305-5