2-Chloro-4-methoxyphenol
Supplier: Apollo Scientific
Warning
Synonyms: 3-Chlor-4-hydroxyanisole
OR0024-25G OR0024-5G
APOSOR0024-25GEA 29 EUR
APOSOR0024-25G APOSOR0024-5G
2-Chloro-4-methoxyphenol
2-Chloro-4-methoxyphenol
Formula: C₇H₇ClO₂
MW: 158.58 g/mol
Melting Pt: 39…44 °C
Density: 1.153 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00070773
CAS Number: 18113-03-6
EINECS: 242-007-3