5-Chloro-2-nitrobenzamide
Supplier: Apollo Scientific
Warning
OR0028-100G OR0028-25G OR0028-5G
APOSOR0028-100GEA 284 EUR
APOSOR0028-100G APOSOR0028-25G APOSOR0028-5G
5-Chloro-2-nitrobenzamide
5-Chloro-2-nitrobenzamide
Formula: C₇H₅ClN₂O₃
MW: 200.58 g/mol
Melting Pt: 157…160 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00017509
CAS Number: 40763-96-0