2-Chlorothioanisole
Supplier: Apollo Scientific
Warning
Synonyms: 1-Chloro-2-(methylthio)benzene, 2-Chlorophenyl methyl sulphide
OR0032-100G OR0032-10G OR0032-25G
APOSOR0032-100GEA 116 EUR
APOSOR0032-100G APOSOR0032-10G APOSOR0032-25G
2-Chlorothioanisole
2-Chlorothioanisole
Formula: C₇H₇ClS
MW: 158.65 g/mol
Boiling Pt: 239…240 °C
Density: 1.2 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00040124
CAS Number: 17733-22-1