2,5-Dibromoaniline 95%
Supplier: Apollo Scientific
Warning
OR0036-25G OR0036-5G OR0036-1G
APOSOR0036-25GEA 0 EUR
APOSOR0036-25G APOSOR0036-5G APOSOR0036-1G
2,5-Dibromoaniline 95%
2,5-Dibromoaniline
Formula: C₆H₅Br₂N
MW: 250.92 g/mol
Melting Pt: 51…56 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007636
CAS Number: 3638-73-1
EINECS: 222-865-5