2'-Nitroacetophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: o-Nitroacetophenone, 1-(2-Nitrophenyl)ethanone
OR0043-10G OR0043-25G
APOSOR0043-10GEA 21.9 EUR
APOSOR0043-10G APOSOR0043-25G
2'-Nitroacetophenone 98%
2'-Nitroacetophenone
Formula: C₈H₇NO₃
MW: 165.15 g/mol
Boiling Pt: 159 °C/16mmHg
Melting Pt: 23…27 °C
Density: 1.24 g/cm³ (25 °C)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00007144
CAS Number: 577-59-3
EINECS: 209-414-8