3-Acetylcoumarin
Supplier: Apollo Scientific
Warning
Synonyms: 3-Acetyl-2H-chromen-2-one
OR0090-5G OR0090-1G OR0090-10G
APOSOR0090-5GEA 34.8 EUR
APOSOR0090-5G APOSOR0090-1G APOSOR0090-10G
3-Acetylcoumarin
3-Acetylcoumarin
Formula: C₁₁H₈O₃
MW: 188.18 g/mol
Melting Pt: 119…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00006853
CAS Number: 3949-36-8