4-Bromo-m-toluidine 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Amino-6-bromotoluene, 4-Bromo-3-methylaniline, 5-Amino-2-bromotoluene
OR0103-25G OR0103-100G
APOSOR0103-25GEA 21.9 EUR
APOSOR0103-25G APOSOR0103-100G
4-Bromo-m-toluidine 98%
4-Bromo-m-toluidine
Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 240 °C
Melting Pt: 79…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00007828
CAS Number: 6933-10-4
EINECS: 230-056-3
UN: 2811
ADR: 6.1,III