2-Benzoxazol-2-yl-1-(4-tert-butylphenyl)ethanol 95+%
Supplier: Apollo Scientific
OR0148-1G
OR0148-5G
APOSOR0148-1GEA
163
EUR
APOSOR0148-1G
APOSOR0148-5G
2-Benzoxazol-2-yl-1-(4-tert-butylphenyl)ethanol 95+%
2-Benzoxazol-2-yl-1-(4-tert-butylphenyl)ethanol
Formula:
C₁₉H₂₁NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD03426055 CAS Number: 849021-34-7 |