1-(4-tert-Butylphenyl)-2-(pyrimidin-4-yl)ethanol 95+%
Supplier: Apollo Scientific
|
Warning
|
OR0149-1G
OR0149-5G
OR0149-25G
APOSOR0149-1GEA
163
EUR
APOSOR0149-1G
APOSOR0149-5G
APOSOR0149-25G
1-(4-tert-Butylphenyl)-2-(pyrimidin-4-yl)ethanol 95+%
1-(4-tert-Butylphenyl)-2-(pyrimidin-4-yl)ethanol
Formula:
C₁₆H₂₀N₂O Boiling Pt: 96…98 °C Density: 1.6 g/cm³ (25 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD03426054 CAS Number: 849021-31-4 |