2,6-Dichloropyridine-4-methanol 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 2,6-Dichloro-4-pyridinemethanol, 2,6-Dichloro-4-(hydroxymethyl)pyridine
OR0150-1G OR0150-5G
APOSOR0150-1GEA 26.1 EUR
APOSOR0150-1G APOSOR0150-5G
2,6-Dichloropyridine-4-methanol 95+%
2,6-Dichloropyridine-4-methanol
Formula: C₆H₅Cl₂NO
MW: 178.0160 g/mol
Melting Pt: 127…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00052638
CAS Number: 101990-69-6
UN: 2811
ADR: 6.1,III