2-Chloro-4-nitroaniline 98%
Supplier: Apollo Scientific
|
Warning
|
OR015002-100G
OR015002-25G
OR015002-5G
APOSOR015002-100GEA
109
EUR
APOSOR015002-100G
APOSOR015002-25G
APOSOR015002-5G
2-Chloro-4-nitroaniline 98%
2-Chloro-4-nitroaniline
Formula:
C₆H₅ClN₂O₂ MW: 172.57 g/mol Boiling Pt: 326 Melting Pt: 108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007665 CAS Number: 121-87-9 UN: 2237 ADR: 6.1 III |