4-(2-Hydroxyethyl)thiomorpholine-1,1-dioxide
Supplier: Apollo Scientific
Warning
Synonyms: 4-(2-Hydroxyethyl)thiomorpholine 1,1-Dioxide, 2-(1,1-Dioxothiomorpholino)ethanol
OR015030-25G OR015030-5G
APOSOR015030-25GEA 681 EUR
APOSOR015030-25G APOSOR015030-5G
4-(2-Hydroxyethyl)thiomorpholine-1,1-dioxide
4-(2-Hydroxyethyl)thiomorpholine-1,1-dioxide
Formula: C₆H₁₃NO₃S
MW: 179.23 g/mol
Boiling Pt: 197 °C/ 1mmHg
Melting Pt: 73…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00106415
CAS Number: 26475-62-7