Imidazo[1,2-a]pyrazine 97%
Supplier: Apollo Scientific
Warning
OR01539-25G
APOSOR01539-25GEA 140 EUR
APOSOR01539-25G
Imidazo[1,2-a]pyrazine 97%
Imidazo[1,2-a]pyrazine
Formula: C₆H₅N₃
MW: 119.1200 g/mol
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD06245371
CAS Number: 274-79-3