2,6-Dichloropyridine-4-carboxaldehyde 96%
Supplier: Apollo Scientific
Warning
Synonyms: 2,6-Dichloroisonicotinaldehyde
OR01602-1G OR01602-5G OR01602-10G
APOSOR01602-1GEA 79.9 EUR
APOSOR01602-1G APOSOR01602-5G APOSOR01602-10G
2,6-Dichloropyridine-4-carboxaldehyde 96%
2,6-Dichloropyridine-4-carboxaldehyde
Formula: C₆H₃Cl₂NO
MW: 176 g/mol
Melting Pt: 44…48 °C
Storage Temperature: Ambient
MDL Number: MFCD00124816
CAS Number: 113293-70-2