Propagyl bromide 80% in toluene
Supplier: Apollo Scientific
Danger
Synonyms: 3-Bromopropyne
OR01606-250ML OR01606-50ML
APOSOR01606-250MLEA 435 EUR
APOSOR01606-250ML APOSOR01606-50ML
Propagyl bromide 80% in toluene
Propagyl bromide
Formula: C₃H₃Br
MW: 118.96 g/mol
Boiling Pt: 88…90 °C
Melting Pt: –61 °C
Density: 1.335 g/cm³ (25 °C)
Flash Pt: 18 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000241
CAS Number: 106-96-7
EINECS: 203-447-1
UN: 2345
ADR: 3,II