2-(1-Hydroxyethyl)pyridine
Supplier: Apollo Scientific
Warning
Synonyms: 1-(Pyridin-2-yl)ethanol
OR01623-1G OR01623-250MG OR01623-5G
APOSOR01623-1GEA 29 EUR
APOSOR01623-1G APOSOR01623-250MG APOSOR01623-5G
2-(1-Hydroxyethyl)pyridine
2-(1-Hydroxyethyl)pyridine
Formula: C₇H₉NO
MW: 123.1525 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06245419
CAS Number: 18728-61-5