Tetrabutylammonium chloride
Supplier: Apollo Scientific
Warning
OR01703-100G OR01703-1KG OR01703-500G
APOSOR01703-100GEA 43.5 EUR
APOSOR01703-100G APOSOR01703-1KG APOSOR01703-500G
Tetrabutylammonium chloride
Tetrabutylammonium chloride
Formula: C₁₆H₃₆ClN
Melting Pt: 70 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00011635
CAS Number: 1112-67-0
EINECS: 214-195-7