tert-Butyl-N-[(5-mercapto-4-methyl-4H-1,2,4-triazol-3-yl)methyl]carbamate
Supplier: Apollo Scientific
Synonyms: 5-(Aminomethyl)-4-methyl-4H-1,2,4-triazole-3-thiol, 5-BOC protected
OR017082-1G OR017082-10G
APOSOR017082-1GEA 111 EUR
APOSOR017082-1G APOSOR017082-10G
tert-Butyl-N-[(5-mercapto-4-methyl-4H-1,2,4-triazol-3-yl)methyl]carbamate
tert-Butyl-N-[(5-mercapto-4-methyl-4H-1,2,4-triazol-3-yl)methyl]carbamate
Formula: C₉H₁₆N₄O₂S
MW: 244.32 g/mol
Melting Pt: 169…172 °C
Storage Temperature: Ambient
MDL Number: MFCD01570587
CAS Number: 519056-65-6