Methyl-5-phenyl-1H-pyrazole-3-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: 3-(Methoxycarbonyl)-5-phenyl-1H-pyrazole, [3-(Methoxycarbonyl)-1H-pyrazol-5-yl]benzene
OR01773-1G OR01773-500MG
APOSOR01773-1GEA 24.8 EUR
APOSOR01773-1G APOSOR01773-500MG
Methyl-5-phenyl-1H-pyrazole-3-carboxylate
Methyl-5-phenyl-1H-pyrazole-3-carboxylate
Formula: C₁₁H₁₀N₂O₂
MW: 202.21 g/mol
Melting Pt: 180…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00781977
CAS Number: 56426-35-8