5-Bromo-2-methox-6-methylypyridine
Supplier: Apollo Scientific
Danger
Synonyms: 3-Bromo-6-methoxy-2-picoline, 3-Bromo-6-methoxy-2-methylpyridine
OR01795-1G OR01795-5G
APOSOR01795-1GEA 21.9 EUR
APOSOR01795-1G APOSOR01795-5G
5-Bromo-2-methox-6-methylypyridine
5-Bromo-2-methox-6-methylypyridine
Formula: C₇H₈BrNO
MW: 202.05 g/mol
Boiling Pt: 212 °C
Density: 1.468 g/cm³ (25 °C)
Flash Pt: 106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094943
CAS Number: 126717-59-7