6-Bromonicotinic acid
Supplier: Apollo Scientific
|
Warning
|
OR0182-25G
OR0182-5G
OR0182-100G
APOSOR0182-25GEA
78.3
EUR
APOSOR0182-25G
APOSOR0182-5G
APOSOR0182-100G
6-Bromonicotinic acid
6-Bromonicotinic acid
Formula:
C₆H₄BrNO₂ Melting Pt: 179…188 °C Storage Temperature: Ambient |
MDL Number:
MFCD01927100 CAS Number: 6311-35-9 |