(±)-3-Butyne-2-ol
Supplier: Apollo Scientific
Danger
Synonyms: (±)-But-3-yn-2-ol, (±)-3-Hydroxy-1-butyne
OR01834-25G OR01834-1G OR01834-5G OR01834-10G OR01834-50G
APOSOR01834-25GEA 101 EUR
APOSOR01834-25G APOSOR01834-1G APOSOR01834-5G APOSOR01834-10G APOSOR01834-50G
(±)-3-Butyne-2-ol
(±)-3-Butyne-2-ol
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 108…110 °C
Density: 0.894 g/cm³ (25 °C)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00004541
CAS Number: 2028-63-9
EINECS: 217-978-1
UN: 2929
ADR: 6.1+3,II