N-Boc-4-bromo-2-methylaniline
Supplier: Apollo Scientific
Synonyms: tert-Butyl (4-bromo-2-methylphenyl)carbamate, tert-Butyl 4-bromo-2-methylphenylcarbamate, 4-Bromo-2-methylaniline, N-BOC protected
OR018874-25G OR018874-5G
APOSOR018874-25GEA 179 EUR
APOSOR018874-25G APOSOR018874-5G
N-Boc-4-bromo-2-methylaniline
N-Boc-4-bromo-2-methylaniline
Formula: C₁₂H₁₆BrNO₂
MW: 286.1700 g/mol
Melting Pt: 159…162 °C
Storage Temperature: Ambient
MDL Number: MFCD01765429
CAS Number: 306937-14-4