4-Chloro-1-methyl-1H-pyrazolo[3,4-d]pyrimidine
Supplier: Apollo Scientific
Danger
OR019232-1G OR019232-250MG
APOSOR019232-1GEA 42.2 EUR
APOSOR019232-1G APOSOR019232-250MG
4-Chloro-1-methyl-1H-pyrazolo[3,4-d]pyrimidine
4-Chloro-1-methyl-1H-pyrazolo[3,4-d]pyrimidine
Formula: C₆H₅ClN₄
MW: 168.5837 g/mol
Melting Pt: 97…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00661967
CAS Number: 23000-43-3
UN: 2811
ADR: 6.1,III