Ethyl-1,3-dimethylpyrazole-5-carboxylate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 1,3-dimethyl-1H-pyrazole-5-carboxylate, 1,3-Dimethylpyrazole-5-carboxylic Acid Ethyl Ester
OR019341-5G OR019341-100G OR019341-25G
APOSOR019341-5GEA 21.9 EUR
APOSOR019341-5G APOSOR019341-100G APOSOR019341-25G
Ethyl-1,3-dimethylpyrazole-5-carboxylate 97%
Ethyl-1,3-dimethylpyrazole-5-carboxylate
Formula: C₈H₁₂N₂O₂
MW: 168.20 g/mol
Boiling Pt: 90 °C
Density: 1.07 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00233452
CAS Number: 5744-40-1