Methyl-3-aminosalicylate
Supplier: Apollo Scientific
Warning
OR01954-1G OR01954-250MG
APOSOR01954-1GEA 62.5 EUR
APOSOR01954-1G APOSOR01954-250MG
Methyl-3-aminosalicylate
Methyl 3-aminosalicylate
Formula: C₈H₉NO₃
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD09842614
CAS Number: 35748-34-6