2-(Chloromethyl)-5-methoxy-1,3-benzoxazole
Supplier: Apollo Scientific
Warning
OR01967-1G OR01967-250MG
APOSOR01967-1GEA 314 EUR
APOSOR01967-1G APOSOR01967-250MG
2-(Chloromethyl)-5-methoxy-1,3-benzoxazole
2-(Chloromethyl)-5-methoxy-1,3-benzoxazole
Formula: C₉H₈ClNO₂
MW: 197.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09836173
CAS Number: 63842-21-7