Ethyl-4,6-dichloro-5-nitronicotinate
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 4,6-dichloro-5-nitronicotinate
OR01969-1G OR01969-250MG
APOSOR01969-1GEA 119 EUR
APOSOR01969-1G APOSOR01969-250MG
Ethyl-4,6-dichloro-5-nitronicotinate
Ethyl-4,6-dichloro-5-nitronicotinate
Formula: C₈H₆Cl₂N₂O₄
MW: 265.05 g/mol
Melting Pt: 36…38.5 °C
Storage Temperature: Ambient
MDL Number: MFCD09842620
CAS Number: 154012-15-4