(±)-2-Phenylpiperazine 97%
Supplier: Apollo Scientific
Warning
OR0220-1G OR0220-5G
APOSOR0220-1GEA 34.8 EUR
APOSOR0220-1G APOSOR0220-5G
(±)-2-Phenylpiperazine 97%
(±)-2-Phenylpiperazine
Formula: C₁₀H₁₄N₂
MW: 162.23 g/mol
Melting Pt: 83…87 °C
Storage Temperature: Ambient
MDL Number: MFCD01871362
CAS Number: 5271-26-1