4-Chloro-3-nitrobenzenesulphonyl chloride 98%

Supplier: Apollo Scientific
Danger

APOSOR0266-1GEA 14.7 EUR
APOSOR0266-1G
4-Chloro-3-nitrobenzenesulphonyl chloride 98%
4-Chloro-3-nitrobenzenesulphonyl chloride
Formula: C₆H₃Cl₂NO₄S
MW: 256,07 g/mol
Melting Pt: 59…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00007440
EINECS: 202-558-2
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR