ω-Bromo-3-chloroacetophenone 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Bromo-3'-chloroacetophenone, Bromomethyl m-chlorophenyl ketone, 2-Bromo-1-(3-chlorophenyl)ethanone, 3-Chlorophenacyl bromide, α-Bromo-3-chloroacetophenone
OR0271-1G OR0271-25G OR0271-5G
APOSOR0271-1GEA 21.9 EUR
APOSOR0271-1G APOSOR0271-25G APOSOR0271-5G
ω-Bromo-3-chloroacetophenone 97%
ω-Bromo-3-chloroacetophenone
Formula: C₈H₆BrClO
MW: 233.49 g/mol
Melting Pt: 37…40 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00792669
CAS Number: 41011-01-2
UN: 1759
ADR: 8,III