3-Chlorothiophene
Supplier: Apollo Scientific
Danger
OR0302-25G OR0302-5G OR0302-100G
APOSOR0302-25GEA 81.2 EUR
APOSOR0302-25G APOSOR0302-5G APOSOR0302-100G
3-Chlorothiophene
3-Chlorothiophene
Formula: C₄H₃ClS
MW: 118.59 g/mol
Boiling Pt: 137…139 °C
Density: 1.276 g/cm³ (25 °C)
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00043887
CAS Number: 17249-80-8
UN: 1993
ADR: 3,III