2'-Deoxyuridine
Supplier: Apollo Scientific
Warning
OR0326-100G OR0326-25G OR0326-5G
APOSOR0326-100GEA 217 EUR
APOSOR0326-100G APOSOR0326-25G APOSOR0326-5G
2'-Deoxyuridine
2'-Deoxyuridine
Formula: C₉H₁₂N₂O₅
MW: 228.20 g/mol
Melting Pt: 165…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00006527
CAS Number: 951-78-0
EINECS: 213-455-7