3,5-Dichlorobenzoyl chloride 98%
Supplier: Apollo Scientific
Danger
OR0343-100G OR0343-25G OR0343-500G
APOSOR0343-100GEA 42.2 EUR
APOSOR0343-100G APOSOR0343-25G APOSOR0343-500G
3,5-Dichlorobenzoyl chloride 98%
3,5-Dichlorobenzoyl chloride
Formula: C₇H₃Cl₃O
Boiling Pt: 245 °C/25mmHg
Melting Pt: 24…28 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00009817
CAS Number: 2905-62-6
EINECS: 220-813-6
UN: 3261
ADR: 8,II