3-Amino-4-pyrazolecarbonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 5-Aminopyrazole-4-carbonitrile, 3-Amino-1H-pyrazole-4-carbonitrile, 3-Aminopyrazole-4-carbonitrile, 3-Amino-4-cyanopyrazole 3-Amino-4-pyrazolecarbonitrile 5-Amino-4-cyanopyrazole 3-Amino-1H-pyrazole-4-carbonitrile, 3-Amino-4-cyanopyrazole
OR0352T-100G OR0352T-25G OR0352T-250G
APOSOR0352T-100GEA 43.5 EUR
APOSOR0352T-100G APOSOR0352T-25G APOSOR0352T-250G
3-Amino-4-pyrazolecarbonitrile
3-Amino-4-pyrazolecarbonitrile
A synthetic intermediate.
Formula: C₄H₄N₄
MW: 108.10 g/mol
Melting Pt: 172…176 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005237
CAS Number: 16617-46-2
EINECS: 240-665-6
UN: 3439
ADR: 6.1 III