2,4-Dichloro-6-methylphenyl isothiocyanate
Supplier: Apollo Scientific
Warning
OR0354-5G OR0354-1G
APOSOR0354-5GEA 418 EUR
APOSOR0354-5G APOSOR0354-1G
2,4-Dichloro-6-methylphenyl isothiocyanate
2,4-Dichloro-6-methylphenyl isothiocyanate
Formula: C₈H₅Cl₂NS
Melting Pt: 57…58 °C
Storage Temperature: Ambient
CAS Number: 306935-83-1