3-(4,4-Dimethyl-2-oxazolinyl)pyridine
Supplier: Apollo Scientific
Warning
Synonyms: 3-(4,5-Dihydro-4,4-dimethyl-1,3-oxazol-2-yl)pyridine, 4,4-Dimethyl-2-(pyridin-3-yl)-4,5-dihydrooxazole
OR0380-1G OR0380-5G OR0380-10G
APOSOR0380-1GEA 58 EUR
APOSOR0380-1G APOSOR0380-5G APOSOR0380-10G
3-(4,4-Dimethyl-2-oxazolinyl)pyridine
3-(4,4-Dimethyl-2-oxazolinyl)pyridine
Formula: C₁₀H₁₂N₂O
MW: 176.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01313220
CAS Number: 68981-86-2