5-Nitroindole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Nitro-1H-indole
OR0385-100G OR0385-25G
APOSOR0385-100GEA 94.4 EUR
APOSOR0385-100G APOSOR0385-25G
5-Nitroindole 98%
5-Nitroindole
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 139…143 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005673
CAS Number: 6146-52-7
EINECS: 228-153-0