4-(Dimethylamino)phenyl isothiocyanate
Supplier: Apollo Scientific
Danger
OR0411-1G OR0411-5G OR0411-25G
APOSOR0411-1GEA 71.2 EUR
APOSOR0411-1G APOSOR0411-5G APOSOR0411-25G
4-(Dimethylamino)phenyl isothiocyanate
4-(Dimethylamino)phenyl isothiocyanate
Formula: C₉H₁₀N₂S
MW: 178.25 g/mol
Melting Pt: 64…66 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041093
CAS Number: 2131-64-8
EINECS: 218-360-4
UN: 2928
ADR: 6.1+8 II