1,2-Dimethylimidazole-4-sulphonyl chloride 95%
Supplier: Apollo Scientific
Danger
Synonyms: 1,2-Dimethyl-1H-imidazole-4-sulphonyl chloride
OR0412-1G OR0412-250MG
APOSOR0412-1GEA 72.5 EUR
APOSOR0412-1G APOSOR0412-250MG
1,2-Dimethylimidazole-4-sulphonyl chloride 95%
1,2-Dimethylimidazole-4-sulphonyl chloride
Formula: C₅H₇ClN₂O₂S
Melting Pt: 91…94 °C
Storage Temperature: Ambient
MDL Number: MFCD01313780
CAS Number: 137049-02-6
UN: 1759
ADR: 8 III